missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxypyrrolidine, 96%

Product Code. 11424730
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424730 1 g 1g
11414730 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424730 Supplier Thermo Scientific Alfa Aesar Supplier No. H50769.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 420137-14-0 | C11H15NO | 177.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420137-14-0
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD08060975
InChI Key CWBMYKUPMLRKQK-UHFFFAOYSA-N
Synonym 3-benzyloxy pyrrolidine, 3-benzyloxypyrrolidine, 3-benzyloxy-pyrrolidine, 3-phenylmethoxy pyrrolidine, benzyl pyrrolidin-3-yl ether, pyrrolidin-3-yl oxy methyl benzene
PubChem CID 14497353
IUPAC Name 3-phenylmethoxypyrrolidine
SMILES C1CNCC1OCC2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 177.25
Percent Purity 96%
Chemical Name or Material 3-Benzyloxypyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.