missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(Boc-amino)-4-methylbenzeneboronic acid, 98%

Product Code. 15499895
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499895 250 mg 250mg
15449287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499895 Supplier Alfa Aesar™ Supplier No. H52548.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 850568-81-9
Molecular Formula C12H18BNO4
Molecular Weight (g/mol) 251.09
MDL Number MFCD04115654
InChI Key RUPLVISWFCBMCR-UHFFFAOYSA-N
Synonym 3-boc-amino-4-methylphenyl boronic acid, 3-tert-butoxycarbonyl amino-4-methylphenyl boronic acid, 3-tert-butoxycarbonyl amino-4-methylphenylboronic acid, 3-boc-amino-4-methylbenzeneboronic acid, 4-methyl-3-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 3-boc-amino-4-methylphenyl boronicacid, 3-t-butoxycarbonylamino-4-methylphenylboronic acid, 3-tert-butoxy carbonyl amino-4-methylphenyl boronic acid, 3-tert-butoxycarbonylamino-4-methylphenylboronic acid
PubChem CID 5173718
IUPAC Name [4-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
SMILES CC1=C(NC(=O)OC(C)(C)C)C=C(C=C1)B(O)O

Specifications

Melting Point 300°C
Quantity 250 mg
Formula Weight 251.09
Percent Purity 98%
Chemical Name or Material 3-(Boc-amino)-4-methylbenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.