missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,4,6-trimethylaniline, 98+%

Product Code. 11304344
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304344 10 g 10g
11314344 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304344 Supplier Thermo Scientific Alfa Aesar Supplier No. L00873.09
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 82842-52-2 | C9H12BrN | 214.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82842-52-2
Molecular Formula C9H12BrN
Molecular Weight (g/mol) 214.106
MDL Number MFCD00015476
InChI Key MVLMPTBHZPYDBZ-UHFFFAOYSA-N
Synonym benzenamine, 3-bromo-2,4,6-trimethyl, pubchem4363, ksc496e8h, 2,4,6-trimethyl-3-bromoaniline, 3-bromo-2,4,6-trimethyl-aniline, 3-bromo-2,4,6-trimethylbenzenamine, 3-bromo-2,4,6-trimethylphenylamine, 3-bromo-2,4,6-trimethyl-phenylamine, 3-bromo-2,4,6-trimethylaniline, 3-bromo-2,4,6-trimethylaniline, 98+%
PubChem CID 688300
IUPAC Name 3-bromo-2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1N)C)Br)C

Specifications

Melting Point 34°C to 38°C
Boiling Point 153°C to 155°C (17 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2829829
Sensitivity Light sensitive
Formula Weight 214.11
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-2,4,6-trimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.