missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,5-dichlorothiophene, 97%

Product Code. 11463284
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463284 1 g 1g
11473284 5 g 5g
11483284 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463284 Supplier Thermo Scientific Alfa Aesar Supplier No. A10603.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 60404-18-4 | C4HBrCl2S | 231.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60404-18-4
Molecular Formula C4HBrCl2S
Molecular Weight (g/mol) 231.92
MDL Number MFCD00041440
InChI Key PBUHOXBSIQJRNO-UHFFFAOYSA-N
PubChem CID 521925
IUPAC Name 3-bromo-2,5-dichlorothiophene
SMILES ClC1=CC(Br)=C(Cl)S1

Specifications

Boiling Point 95°C to 97°C (14 mmHg)
Refractive Index 1.608
Quantity 1 g
Beilstein 1306272
Synonym thiophene,3-bromo-2,5-dichloro, zlchem 1326, acmc-209mjs, 2,5-dichloro bromo thiophene, 2,5-dichloro-3-bromothiophene, 3-bromo-2,5-dichloro thiophene, thiophene, 3-bromo-2,5-dichloro
Formula Weight 231.93
Percent Purity 97%
Chemical Name or Material 3-Bromo-2,5-dichlorothiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.