missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Product Code. 15414927
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15414927

Brand: Thermo Scientific Alfa Aesar H32228.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149947-16-0
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD09842442
InChI Key MGVHFTWDCYIBDO-UHFFFAOYSA-N
PubChem CID 18989440
IUPAC Name 1-bromo-3-(bromomethyl)-2-fluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)CBr

Specifications

Boiling Point 60°C (0.1 mmHg)
Quantity 5 g
UN Number UN3265
Synonym 1-bromo-3-bromomethyl-2-fluorobenzene, 3-bromo-2-fluorobenzyl bromide, benzene,1-bromo-3-bromomethyl-2-fluoro, acmc-20ahso, intermediates-zcf02146, benzene, 1-bromo-3-bromomethyl-2-fluoro
Formula Weight 267.92
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-fluorobenzyl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.