missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylaniline, 98+%

Product Code. p-7081701
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11453854

Brand: Thermo Scientific Alfa Aesar A11884.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55289-36-6 | C7H8BrN | 186.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55289-36-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD00051579
InChI Key IILVSKMKMOJHMA-UHFFFAOYSA-N
Synonym 2-amino-6-bromotoluene, 3-bromo-o-toluidine, benzenamine, 3-bromo-2-methyl, 3-bromo-2-methylbenzenamine, 2-bromo-6-aminotoluene, 3-bromo-2-methyl-phenylamine, 3-bromo-2-methyl-aniline, 3-bromo-2-methylphenylamine, 3-bromo-2-methyl-benzenamine, 3-brom-2-methylanilin
PubChem CID 123538
IUPAC Name 3-bromo-2-methylaniline
SMILES CC1=C(C=CC=C1Br)N

Specifications

Density 1.51
Boiling Point 253°C to 255°C
Flash Point >110°C (230°F)
Refractive Index 1.616
Quantity 5 g
UN Number UN2810
Beilstein 2354213
Sensitivity Light sensitive
Formula Weight 186.06
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-2-methylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.