missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylbenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11396709
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11396709

Brand: Thermo Scientific Alfa Aesar H26249.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Bromo-2-methylbenzotrifluoride is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69902-83-6
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.035
MDL Number MFCD04038231
InChI Key OYFKTZQGAWXNTI-UHFFFAOYSA-N
Synonym 1-bromo-2-methyl-3-trifluoromethyl benzene, 3-bromo-2-methylbenzotrifluoride, 2-methyl-3-trifluoromethyl bromobenzene, 2-bromo-6-trifluoromethyl toluene, 2-bromo-6-trifluoromethyltoluene, acmc-209obm, ksc495a6l, 3-bromo-2-methyltrifluoromethylbenzene, 3-bromo-2-methyl trifluoromethylbenzene
PubChem CID 2775634
IUPAC Name 1-bromo-2-methyl-3-(trifluoromethyl)benzene
SMILES CC1=C(C=CC=C1Br)C(F)(F)F

Specifications

Boiling Point 35°C (1 mmHg)
Quantity 1 g
Solubility Information Not miscible or difficult to mix.
Formula Weight 239.04
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-methylbenzotrifluoride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.