missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorophenol, 98%, Thermo Scientific Chemicals

Product Code. 15468268
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468268 5 g 5g
15458268 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468268 Supplier Thermo Scientific Alfa Aesar Supplier No. H61702.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27407-11-0
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD03425884
InChI Key QWTULQLVGNZMLF-UHFFFAOYSA-N
Synonym 3-bromo-4-fluoro-phenol, phenol, 3-bromo-4-fluoro, pubchem1486, acmc-1cmha, ksc201s9b, 3-bromanyl-4-fluoranyl-phenol
PubChem CID 2783381
IUPAC Name 3-bromo-4-fluorophenol
SMILES C1=CC(=C(C=C1O)Br)F

Specifications

Color Yellow to Green
Quantity 5 g
UN Number UN2811
Formula Weight 191
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 3-Bromo-4-fluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.