missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxybenzonitrile, 98%

Product Code. p-7024548
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11473997

Brand: Thermo Scientific Alfa Aesar A19400.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2315-86-8 | C7H4BrNO | 198.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2315-86-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00143096
InChI Key HLHNOIAOWQFNGW-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r
PubChem CID 75341
IUPAC Name 3-bromo-4-hydroxybenzonitrile
SMILES OC1=CC=C(C=C1Br)C#N

Specifications

Melting Point 156°C to 161°C
Quantity 5 g
UN Number UN3439
Beilstein 2207020
Formula Weight 198.02
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-hydroxybenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.