missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybenzoic acid, 98+%

Product Code. 11410513
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410513 5 g 5g
11420513 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410513 Supplier Thermo Scientific Alfa Aesar Supplier No. A12653.06
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 99-58-1 | C8H6BrO3 | 230.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-58-1
Molecular Formula C8H6BrO3
Molecular Weight (g/mol) 230.04
MDL Number MFCD00020295
InChI Key BBPZABXVRBFWGD-UHFFFAOYSA-M
Synonym 3-bromo-p-anisic acid, benzoic acid, 3-bromo-4-methoxy, 3-bromo-4-methoxybenzoicacid, akos bbb/210, rarechem al bo 0751, timtec-bb sbb009955, 4-methoxy-3-bromo benzoic acid, 3-bromanissaure, pubchem2675, 2-bromo-4-carboxyanisole
PubChem CID 66836
IUPAC Name 3-bromo-4-methoxybenzoic acid
SMILES COC1=CC=C(C=C1Br)C([O-])=O

Specifications

Melting Point 217°C to 222°C
Odor Odorless
Quantity 5 g
Beilstein 2087585
Formula Weight 231.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-methoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.