missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylaniline, 97%

Product Code. 11450528
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450528 5 g 5g
11460528 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450528 Supplier Thermo Scientific Alfa Aesar Supplier No. B24670.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7745-91-7 | C7H8BrN | 186.05 g/mol

3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger?s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger′s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7745-91-7
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00134176
InChI Key GRXMMIBZRMKADT-UHFFFAOYSA-N
Synonym 3-bromo-p-toluidine, p-toluidine, 3-bromo, benzenamine, 3-bromo-4-methyl, 3-bromo-4-methyl-phenylamine, 3-bromo-1,4-toluidine, 4-amino-2-bromotoluene, 3-bromo-4-methylbenzenamine, 3-bromo-4-methylphenylamine, 3-bromo-para-toluidine, 3-bromo-4-methyl-aniline
PubChem CID 82187
IUPAC Name 3-bromo-4-methylaniline
SMILES CC1=CC=C(N)C=C1Br

Specifications

Melting Point 25°C to 28°C
Boiling Point 254°C to 257°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.6115
Quantity 5 g
UN Number UN2811
Beilstein 1562057
Solubility Information Insoluble in water.
Formula Weight 186.06
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.