missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylaniline, 97%, Thermo Scientific™

Product Code. 10460242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10460242 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10460242 Supplier Thermo Scientific Acros Supplier No. 339110050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7745-91-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 27°C to 30°C
Boiling Point 254°C to 257°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8BrN
Linear Formula BrC6H3(CH3)NH2
Refractive Index 1.6110 to 1.613
MDL Number MFCD00134176
Quantity 5 g
Synonym 3-bromo-p-toluidine, p-toluidine, 3-bromo, benzenamine, 3-bromo-4-methyl, 3-bromo-4-methyl-phenylamine, 3-bromo-1,4-toluidine, 4-amino-2-bromotoluene, 3-bromo-4-methylbenzenamine, 3-bromo-4-methylphenylamine, 3-bromo-para-toluidine, 3-bromo-4-methyl-aniline
InChI Key GRXMMIBZRMKADT-UHFFFAOYSA-N
SMILES CC1=CC=C(N)C=C1Br
IUPAC Name 3-bromo-4-methylaniline
Molecular Weight (g/mol) 186.05
PubChem CID 82187
Formula Weight 186.05
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 3-Bromo-4-methylaniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-methylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.