missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylbenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11984551 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
5g
This item is not returnable. View return policy

Product Code. 11984551

Brand: Thermo Scientific Acros 446680050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 66417-30-9
Boiling Point 88.0°C (31.0 Torr)
Assay Percent Range 96% min. (GC)
Molecular Formula C8H6BrF3
MDL Number MFCD01631584
Quantity 5 g
Synonym 2-bromo-1-methyl-4-trifluoromethyl benzene, 3-bromo-4-methylbenzotrifluoride, 2-methyl-5-trifluoromethyl bromobenzene, 2-bromo-4-trifluoromethyl toluene, benzene, 2-bromo-1-methyl-4-trifluoromethyl, 2-bromo-1-methyl-4-trifluoromethyl-benzene, acmc-209nvg, ksc294o7j, 2-bromo-4-trifluoromethyltoluene
InChI Key TWWUWLMGYSLODU-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1Br)C(F)(F)F
IUPAC Name 2-bromo-1-methyl-4-(trifluoromethyl)benzene
Molecular Weight (g/mol) 239.04
PubChem CID 2779376
Formula Weight 239.03
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methylbenzotrifluoride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-methylbenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.