missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylphenol, 98%

Product Code. 11452110
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452110 1 g 1g
15443827 250 mg 250mg
11462110 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452110 Supplier Thermo Scientific Alfa Aesar Supplier No. H28689.03
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 60710-39-6 | C7H7BrO | 187.036 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60710-39-6
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD08273793
InChI Key GMZKNRDHSHYMHG-UHFFFAOYSA-N
Synonym 3-bromo-p-cresol, 2-bromo-4-hydroxytoluene, phenol, 3-bromo-4-methyl, 3-bromo-4-methyl-phenol, 3-brom-p-kresol, acmc-209mlm, 3-bromo-4-methyl phenol, phenol,3-bromo-4-methyl, 3-bromanyl-4-methyl-phenol, ksc494c1p
PubChem CID 10307913
IUPAC Name 3-bromo-4-methylphenol
SMILES CC1=C(C=C(C=C1)O)Br

Specifications

Melting Point 54°C to 57°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 187.04
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-methylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.