missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chlorophenol, ≥97%, Thermo Scientific Chemicals

Product Code. 15422239
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15422239

Brand: Thermo Scientific Alfa Aesar H64844.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56962-04-0
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.45
MDL Number MFCD07780676
InChI Key GMGWXLPFRHYWAS-UHFFFAOYSA-N
Synonym phenol, 3-bromo-5-chloro, 3-bromo-5-chloro-phenol, 3-bromo-5-chlorophenol, 97+%, pubchem17565, acmc-209lv4, ksc268s7h
PubChem CID 22630180
IUPAC Name 3-bromo-5-chlorophenol
SMILES OC1=CC(Cl)=CC(Br)=C1

Specifications

Melting Point 66°C to 70°C
Quantity 25 g
Beilstein 3234780
Formula Weight 207.45
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 3-Bromo-5-chlorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.