missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11439176
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11439176

Brand: Thermo Scientific Alfa Aesar A18034.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130723-13-6
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD00042498
InChI Key LIGBGEJPUQBLTG-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorobenzotrifluoride, 1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluorotrifluoromethylbenzene, 3-fluoro-5-trifluoromethyl bromobenzene, 5-fluoro-3-bromobenzotrifluoride, 3-fluoro-5-trifluoromethylbromobenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-bromo-5-fluoro-trifluoromethylbenzene, benzene, 1-bromo-3-fluoro-5-trifluoromethyl
PubChem CID 2736323
IUPAC Name 1-bromo-3-fluoro-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1F)Br)C(F)(F)F

Specifications

Density 1.511
Boiling Point 146°C
Flash Point >110°C (230°F)
Refractive Index 1.456
Quantity 1 g
Formula Weight 243
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-fluorobenzotrifluoride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.