missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 10843381 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10843381

Brand: Thermo Scientific Acros 435500010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 130723-13-6
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243
MDL Number MFCD00042498
InChI Key LIGBGEJPUQBLTG-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorobenzotrifluoride, 1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluorotrifluoromethylbenzene, 3-fluoro-5-trifluoromethyl bromobenzene, 5-fluoro-3-bromobenzotrifluoride, 3-fluoro-5-trifluoromethylbromobenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-bromo-5-fluoro-trifluoromethylbenzene, benzene, 1-bromo-3-fluoro-5-trifluoromethyl
PubChem CID 2736323
IUPAC Name 1-bromo-3-fluoro-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1F)Br)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5100g/mL
Boiling Point 146°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.51
Formula Weight 243
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3-Bromo-5-fluorobenzotrifluoride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-5-fluorobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.