missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorophenol, 97%

Product Code. p-7036854
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15461498

Brand: Thermo Scientific Alfa Aesar H55288.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 433939-27-6 | C6H4BrFO | 190.999 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433939-27-6
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD07783710
InChI Key JCPJGUPQZDEZQH-UHFFFAOYSA-N
Synonym 3-fluoro-5-bromophenol, 5-bromo-3-fluorophenol, 3-bromo-5-fluoro-phenol, phenol, 3-bromo-5-fluoro, pubchem4129, acmc-1apwb, ksc493m6t, 3-bromo-5-fluorophenol
PubChem CID 21904636
IUPAC Name 3-bromo-5-fluorophenol
SMILES C1=C(C=C(C=C1F)Br)O

Specifications

Melting Point 36°C to 40°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-fluorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.