missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorotoluene, 98%

Product Code. 15418108
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418108 100 g 100g
15408108 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418108 Supplier Thermo Scientific Alfa Aesar Supplier No. H61544.22
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 202865-83-6 | C7H6BrF | 189.027 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-83-6
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD01861195
InChI Key CTNFNUQREIIROB-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorotoluene, 3-fluoro-5-bromotoluene, 5-bromo-3-fluorotoluene, benzene, 1-bromo-3-fluoro-5-methyl, 1-bromo-3-fluoro-5-methyl-benzene, 3-bromo-5-fluoro toluene, pubchem3237, acmc-1cod9, ksc493k3l, tpc-i111
PubChem CID 2773396
IUPAC Name 1-bromo-3-fluoro-5-methylbenzene
SMILES CC1=CC(=CC(=C1)Br)F

Specifications

Color Colorless
Quantity 100 g
Formula Weight 189.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-fluorotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.