missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-iodobenzoic acid, 97%

Product Code. p-7023482
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408946 5 g 5g
11418946 25 g 25g
11428946 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408946 Supplier Thermo Scientific Alfa Aesar Supplier No. A17076.06
Add to basket
This item is not returnable. View return policy

CAS: 188815-32-9 | C7H4BrIO2 | 326.915 g/mol

3-Bromo-5-iodobenzoic acid may be used in the preparation of the following:

  • Phenyl(3-bromo-5-iodo)benzoate.
  • As starting reagent for the large-scale synthesis of the thromboxane receptor antagonist 3-{3-[2-(4-chlorobenzenesulfonamido)ethyl]-5-(4-fluorobenzyl)phenyl}propionic acid, via regioselective Heck cross-coupling reaction.
  • Methyl 3-bromo-5-iodobenzoate.
  • 3-Bromo-5-(triisopropylsilylethynyl)benzoic acid, via Sonogashira coupling reaction.
  • Trifluoroacetophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-iodobenzoic acid may be used in the preparation of the following: • Phenyl(3-bromo-5-iodo)benzoate. • As starting reagent for the large-scale synthesis of the thromboxane receptor antagonist 3-{3-[2-(4-chlorobenzenesulfonamido)ethyl]-5-(4-fluorobenzyl)phenyl}propionic acid, via regioselective Heck cross-coupling reaction. • Methyl 3-bromo-5-iodobenzoate. • 3-Bromo-5-(triisopropylsilylethynyl)benzoic acid, via Sonogashira coupling reaction. • Trifluoroacetophenone.

Solubility
Soluble in methanol. Sparingly Soluble (0.20 g/L) (25°C), Calc.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188815-32-9
Molecular Formula C7H4BrIO2
Molecular Weight (g/mol) 326.915
MDL Number MFCD00191851
InChI Key MKJBJYCBKXPQSY-UHFFFAOYSA-N
Synonym 5-bromo-3-iodobenzoic acid, 3-bromo-5-iodo-benzoic acid, 3-bromo-5-iodobenzoicacid, benzoic acid, 3-bromo-5-iodo, 3-iodo-5-bromobenzoic acid, pubchem3788, acmc-209erv, 3-bromo-5-iodobenzoic aicd, 3-bromo-5-iodo benzoic acid, 5-iodo-3-bromo-benzoic acid
PubChem CID 5182520
IUPAC Name 3-bromo-5-iodobenzoic acid
SMILES C1=C(C=C(C=C1Br)I)C(=O)O

Specifications

Melting Point 218°C to 221°C
Quantity 5 g
Beilstein 3240837
Sensitivity Light sensitive
Solubility Information Soluble in methanol. Sparingly Soluble (0.20g/L) (25°C),Calc.
Formula Weight 326.92
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-iodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.