missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-nitrosalicylaldehyde, 98%

Product Code. p-7058319
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11317126

Brand: Thermo Scientific Alfa Aesar L07071.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16789-84-7 | C7H4BrNO4 | 246.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16789-84-7
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD00051833
InChI Key BESBCGANGAEHPM-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-nitrobenzaldehyde, 3-bromo-5-nitrosalicylaldehyde, 3-bromo-2-hydroxy-5-nitrobenzenecarbaldehyde, 2-hydroxy-3-bromo-5-nitrobenzaldehyde, 3-bromo-2-hydroxy-5-nitro-benzaldehyde, acmc-1cfaz, 3-bromo-5-nitro-salicylaldehyde, benzaldehyde,3-bromo-2-hydroxy-5-nitro, 3-bromanyl-5-nitro-2-oxidanyl-benzaldehyde
PubChem CID 519307
IUPAC Name 3-bromo-2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O

Specifications

Melting Point 147°C to 151°C
Quantity 1 g
Beilstein 1213613
Sensitivity Air Sensitive
Formula Weight 246.02
Percent Purity 98%
Chemical Name or Material 3-Bromo-5-nitrosalicylaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.