missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethoxy)benzonitrile, 97%, Thermo Scientific™

Product Code. 15404417
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404417 1 g 1g
15414417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404417 Supplier Thermo Scientific Alfa Aesar Supplier No. H31778.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914635-52-2
Molecular Formula C8H3BrF3NO
Molecular Weight (g/mol) 266.017
MDL Number MFCD08458074
InChI Key PQPKERBGCBHRJO-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethoxy benzonitrile, 5-bromo-3-trifluoromethoxy benzenecarbonitrile, 3-bromo-5-cyano-alpha,alpha,alpha-trifluoroanisole
PubChem CID 44717376
IUPAC Name 3-bromo-5-(trifluoromethoxy)benzonitrile
SMILES C1=C(C=C(C=C1OC(F)(F)F)Br)C#N

Specifications

Melting Point 35°C to 38°C
Quantity 1 g
UN Number UN3439
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzonitrile
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.