missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)benzene-1,2-diamine, 97%, Thermo Scientific™

Product Code. 10779385
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10779385 1 g 1g
10378522 10 g 10g
10585344 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10779385 Supplier Thermo Scientific Maybridge Supplier No. BTB05134DA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 113170-72-2
Molecular Formula C7H6BrF3N2
Molecular Weight (g/mol) 255.038
MDL Number MFCD00042484
InChI Key OVGAPUVMLZWQQF-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl benzene-1,2-diamine, 3-bromo-4,5-diaminobenzotrifluoride, buttpark 82\03-57, 1,2-diamino-3-bromo-5-trifluoromethyl benzene, 3-bromo-5-trifluoromethyl-1,2-phenylenediamine, 2-amino-3-bromo-5-trifluoromethyl phenylamine, 3-bromo-5-trifluoromethyl-1,2-benzenediamine, 4,5-diamino-3-bromobenzotrifluoride, pubchem2253
PubChem CID 735785
IUPAC Name 3-bromo-5-(trifluoromethyl)benzene-1,2-diamine
SMILES C1=C(C=C(C(=C1N)N)Br)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 255.04
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzene-1, 2-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.