missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9H-carbazole, 97%, Thermo Scientific™

Product Code. 10314292
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10314292 250 mg Amber Glass Bottle 250mg
10502662 1 g Amber Glass Bottle 1g
10632275 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10314292 Supplier Thermo Scientific Maybridge Supplier No. JFD01173CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1592-95-6
Molecular Formula C12H8BrN
Molecular Weight (g/mol) 246.107
InChI Key LTBWKAYPXIIVPC-UHFFFAOYSA-N
Synonym 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc
PubChem CID 252446
IUPAC Name 3-bromo-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 246.11
Percent Purity 97%
Chemical Name or Material 3-Bromo-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.