missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoaniline, 98%

Product Code. 11420264
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420264 25 g 25g
11430264 100 g 100g
11440264 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420264 Supplier Thermo Scientific Alfa Aesar Supplier No. A13665.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 591-19-5 | C6H6BrN | 172.03 g/mol

3-Bromoaniline is used in the synthesis of amino substituted quinazoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoaniline is used in the synthesis of amino substituted quinazoline.

Solubility
Soluble in water (0.97 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-19-5
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00007757
InChI Key DHYHYLGCQVVLOQ-UHFFFAOYSA-N
Synonym m-bromoaniline, benzenamine, 3-bromo, 3-bromo aniline, m-aminobromobenzene, aniline, m-bromo, 3-bromobenzenamine, 3-bromophenylamine, bromoaniline 3-, 3-bromo-benzenamine, 3-bromo-phenylamine
PubChem CID 11562
IUPAC Name 3-bromoaniline
SMILES NC1=CC=CC(Br)=C1

Specifications

Melting Point 16°C
Density 1.580
Boiling Point 251°C
Flash Point >110°C (230°F)
Assay Percent Range 98%
Refractive Index 1.625
Quantity 25 g
UN Number UN2810
Beilstein 742028
Solubility Information Soluble in water (0.97g/L at 20°C).
Formula Weight 172.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromoaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.