missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoanisole, 98+%

Product Code. 11447517
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447517 500 g 500g
11427517 25 g 25g
11437517 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447517 Supplier Thermo Scientific Alfa Aesar Supplier No. A15625.36
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2398-37-0 | C7H7BrO | 187.04 g/mol

3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

Solubility
Miscible in methanol, benzene, hexane, toluene and dichloromethane. Practically insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2398-37-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000081
InChI Key PLDWAJLZAAHOGG-UHFFFAOYSA-N
Synonym 3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
PubChem CID 16971
IUPAC Name 1-bromo-3-methoxybenzene
SMILES COC1=CC=CC(Br)=C1

Specifications

Melting Point ∼2°C
Density 1.489
Boiling Point 210°C to 211°C
Flash Point 93°C (199°F)
Refractive Index 1.564
Quantity 500 g
Beilstein 1524687
Solubility Information Miscible in methanol,benzene,hexane,toluene and dichloromethane. Practically insoluble in water.
Formula Weight 187.04
Percent Purity ≥98%
Chemical Name or Material 3-Bromoanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.