missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzotrifluoride, 98+%

Product Code. 11457756
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457756 25 g 25g
11467756 100 g 100g
11477756 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457756 Supplier Thermo Scientific Alfa Aesar Supplier No. A14724.14
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 401-78-5 | C7H4BrF3 | 225.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-78-5
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00000380
InChI Key NNMBNYHMJRJUBC-UHFFFAOYSA-N
Synonym 3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
PubChem CID 9817
IUPAC Name 1-bromo-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)Br)C(F)(F)F

Specifications

Density 1.613
Boiling Point 151°C to 152°C
Flash Point 59°C (138°F)
Odor Aromatic
Refractive Index 1.473
Quantity 25 g
UN Number UN1993
Beilstein 1449557
Formula Weight 225.01
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzotrifluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Bromobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.