missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoyl chloride, 97%

Product Code. p-4089612 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10603035

Brand: Thermo Scientific Acros 397450050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1711-09-7 | C7H4BrClO | 219.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-09-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD00000669
InChI Key PBOOZQFGWNZNQE-UHFFFAOYSA-N
Synonym m-bromobenzoyl chloride, benzoyl chloride, 3-bromo, 3-bromobenzoylchloride, benzoyl chloride, m-bromo, 3-bromobenzoic acid chloride, attercop-chm at106527, 3-brom-benzoylchlorid, 3-bromo-benzoylchlorid, 3-bromobenzoic chloride, pubchem13574
PubChem CID 74377
IUPAC Name 3-bromobenzoyl chloride
SMILES C1=CC(=CC(=C1)Br)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.6660g/mL
Boiling Point 247°C to 251°C
Flash Point 107°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCl
Quantity 5 g
Specific Gravity 1.666
Formula Weight 219.46
Percent Purity 97%
Chemical Name or Material 3-Bromobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.