missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylamine hydrochloride, 98%

Product Code. 11423177
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423177 5 g 5g
11433177 25 g 25g
11443177 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423177 Supplier Thermo Scientific Alfa Aesar Supplier No. A16132.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39959-54-1 | C7H9BrClN | 222.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39959-54-1
Molecular Formula C7H9BrClN
Molecular Weight (g/mol) 222.51
MDL Number MFCD00012856
InChI Key UGEQUCUBWNAUJS-UHFFFAOYSA-N
Synonym 3-bromobenzylamine hydrochloride, 3-bromophenyl methanamine hydrochloride, 3-bromobenzylamine hcl, 3-bromobenzylaminehydrochloride, m-bromobenzylamine hydrochloride, 3-bromophenyl methylamine hydrochloride, 1-3-bromophenyl methanamine hydrochloride, benzenemethanamine, 3-bromo-, hydrochloride, pubchem3750, 3-bromobenzyl amine hcl
PubChem CID 2724980
IUPAC Name (3-bromophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)Br)CN.Cl

Specifications

Melting Point 218°C to 221°C
Quantity 5 g
Beilstein 3913185
Sensitivity Hygroscopic
Formula Weight 222.52
Percent Purity 98%
Chemical Name or Material 3-Bromobenzylamine hydrochloride
compliance-icons
Product Identifier
  • 3-Bromobenzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 3-Bromobenzylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.