missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10729173
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10729173 1 g Amber Glass Bottle 1g
10664205 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10729173 Supplier Thermo Scientific Maybridge Supplier No. MO07388DA
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 82072-23-9
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
InChI Key OEPGAYXSRGROSQ-UHFFFAOYSA-N
Synonym 3-bromomethyl benzaldehyde, benzaldehyde, 3-bromomethyl, 3-bromomethyl-benzaldehyde, pubchem17017, alpha-bromo-m-tolualdehyde, ksc447q4h
PubChem CID 7127825
IUPAC Name 3-(bromomethyl)benzaldehyde
SMILES C1=CC(=CC(=C1)CBr)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.05
Percent Purity 97%
Chemical Name or Material 3-(Bromomethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.