missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenol, 98%

Product Code. 10103720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging unitSize
10103720 10 g Glass bottle 10g
10113720 25 g Glass bottle 25g
10033663 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10103720 Proveedor Thermo Scientific Acros N.º de proveedor 146340100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 591-20-8 | C6H5BrO | 173.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 591-20-8
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.01
MDL Number MFCD00002253
InChI Key MNOJRWOWILAHAV-UHFFFAOYSA-N
Synonym m-bromophenol, phenol, 3-bromo, 3-bromo phenol, 3-bromo-phenol, phenol, m-bromo, unii-vmu0x6956y, meta bromophenol, meta-bromophenol, 3 bromophenol, 3-bromanylphenol
PubChem CID 11563
IUPAC Name 3-bromophenol
SMILES OC1=CC=CC(Br)=C1

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point 30.0°C to 32.0°C
Color Brown or Yellow
Boiling Point 236.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OH
Refractive Index 1.5950 to 1.5990
Quantity 10 g
Beilstein 06,198
Merck Index 15,1439
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies,alcohol,ether,acetone
Formula Weight 173.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromophenol
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 3-Bromophenol
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.