missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenyl isocyanate, 97%

Product Code. 10382823 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10382823 1 g 1g
10636871 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10382823 Supplier Thermo Scientific Acros Supplier No. 314130010
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 23138-55-8 | C7H4BrNO | 198.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23138-55-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00002014
InChI Key VQVBCZQTXSHJGF-UHFFFAOYSA-N
Synonym 3-bromophenyl isocyanate, 3-bromophenylisocyanate, benzene, 1-bromo-3-isocyanato, isocyanic acid 3-bromophenyl ester, 3-bromobenzenisocyanate, m-bromophenylisocyanate, 3-bromophenyliso-cyanate, 3-bromo phenyl isocyanate, 3-bromo-phenyl isocyanate, acmc-209g1f
PubChem CID 141001
IUPAC Name 1-bromo-3-isocyanatobenzene
SMILES BrC1=CC=CC(=C1)N=C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5860g/mL
Boiling Point 220°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4NCO
Refractive Index 1.5835 to 1.5865
Quantity 1 g
Specific Gravity 1.586
Formula Weight 198.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromophenyl isocyanate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.