missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenylhydrazine hydrochloride, 98%

Product Code. 10217263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217263 25 g Glass bottle 25g
10011950 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217263 Supplier Thermo Scientific Acros Supplier No. 167350250
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 27246-81-7 | C6H8BrClN2 | 223.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27246-81-7
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.50
MDL Number MFCD00012933
InChI Key RPYIPFXHIKXRKS-UHFFFAOYSA-N
Synonym 3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
PubChem CID 33757
IUPAC Name (3-bromophenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC(Br)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 225°C to 231°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrC6H4NHNH2·HCl
Quantity 25 g
Beilstein 15, 433
Formula Weight 223.51
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Bromophenylhydrazine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.