missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotoluene, 98%, Thermo Scientific Chemicals

Product Code. 10215850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215850 100 mL Glass bottle 100mL
10205850 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215850 Supplier Thermo Scientific Acros Supplier No. 173411000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-17-3 | C7H7Br | 171.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-17-3
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000085
InChI Key WJIFKOVZNJTSGO-UHFFFAOYSA-N
Synonym 3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
PubChem CID 11560
IUPAC Name 1-bromo-3-methylbenzene
SMILES CC1=CC=CC(Br)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless to Yellow
Density 1.4100g/mL
Boiling Point 183.0°C to 184.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4Br
Refractive Index 1.5510 to 1.5530
Quantity 100 mL
Beilstein 05, 305
Merck Index 14, 1439
Specific Gravity 1.41
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 171.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Bromotoluene, 99%
Show More Show Less

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.