missing translation for 'onlineSavingsMsg'
Learn More

(3-Carboxypropyl)trimethylammonium chloride, MedChemExpress

Product Code. 16405576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16405576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16405576 Supplier Medchem Express Supplier No. HY113270A
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (3-Carboxypropyl)trimethylammonium chloride is angiopathic substance produced as an intermediary metabolite by gut microbiota that feed on carnitine in dietary red meat.

(3-Carboxypropyl)trimethylammonium chloride is angiopathic substance produced as an intermediary metabolite by gut microbiota that feed on carnitine in dietary red meat.

Specifications

Chemical Name or Material (3-Carboxypropyl)trimethylammonium chloride
Physical Form Solid
CAS 6249-56-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H16ClNO2
For Use With (Application) COVID-19-immunoregulation
Synonym γ-Butyrobetaine hydrochloride
Solubility Information DMSO : 30 mg/mL (165.14 mM; Need ultrasonic)
SMILES C[N+](C)(C)CCCC(O)=O.[Cl-]
Molecular Weight (g/mol) 181.66
Formula Weight 181.66
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.