missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-nitrobenzoic acid, 97%

Product Code. p-7029036
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11405820

Brand: Thermo Scientific Alfa Aesar H26467.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4771-47-5 | C7H4ClNO4 | 201.562 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4771-47-5
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007087
InChI Key VCHSXYHBMFKRBK-UHFFFAOYSA-N
Synonym 3-chloro-2-nitrobenzoic acid, benzoic acid, 3-chloro-2-nitro, 3-chloro-2-nitrobenzoicacid, 3-chloro-2-nitro-benzoic acid, chlornitrobenzoesaure, pubchem11043, chloro-2-nitrobenzoic acid, acmc-209ka6, 3-chloro2-nitrobenzoic acid, ksc237s1n
PubChem CID 20902
SMILES C1=CC(=C(C(=C1)Cl)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 234°C to 240°C
Odor Odorless
Quantity 1 g
Beilstein 2109828
Formula Weight 201.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.