missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-nitrobenzoic acid, 98%, ACROS Organics™

Product Code. 10407370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10407370 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10407370 Supplier Thermo Scientific Acros Supplier No. 151690010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4771-47-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 236°C to 240°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H4ClNO4
Linear Formula ClC6H3(NO2)CO2H
MDL Number MFCD00007087
Quantity 1 g
Synonym 3-chloro-2-nitrobenzoic acid, benzoic acid, 3-chloro-2-nitro, 3-chloro-2-nitrobenzoicacid, 3-chloro-2-nitro-benzoic acid, chlornitrobenzoesaure, pubchem11043, chloro-2-nitrobenzoic acid, acmc-209ka6, 3-chloro2-nitrobenzoic acid, ksc237s1n
InChI Key VCHSXYHBMFKRBK-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)[N+](=O)[O-])C(=O)O
Molecular Weight (g/mol) 201.562
PubChem CID 20902
Formula Weight 201.57
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-nitrobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-2-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.