missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzene-1-thiol, Tech., Thermo Scientific™

Product Code. 10420944
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420944 1 g Amber Glass Bottle 1g
10602665 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420944 Supplier Thermo Scientific Maybridge Supplier No. TL00315DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 60811-23-6
Molecular Formula C6H3ClFS
Molecular Weight (g/mol) 161.60
MDL Number MFCD00052938
InChI Key SFSHSIFYTWIGSF-UHFFFAOYSA-M
Synonym 3-chloro-4-fluorothiophenol, 3-chloro-4-fluorobenzene-1-thiol, 3-chloro-4-fluoro-benzenethiol, pubchem2857, acmc-209wz2, 3-chloro-4-flurothiophenol, benzenethiol, 3-chloro-4-fluoro
PubChem CID 2734197
IUPAC Name 3-chloro-4-fluorobenzenethiol
SMILES FC1=CC=C([S-])C=C1Cl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.61
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Chloro-4-fluorobenzene-1-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.