missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzeneboronic acid, 98%

Product Code. 11449777
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449777 25 g 25g
11429777 1 g 1g
11439777 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449777 Supplier Thermo Scientific Alfa Aesar Supplier No. B22755.14
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 144432-85-9 | C6H5BClFO2 | 174.362 g/mol

3-Chloro-4-fluorobenzeneboronic acid is used as a reactant for Rh-catalyzed asymmetric addition reactions, palladium-catalyzed oxidative cross-coupling reaction and Suzuki-Miyaura coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-4-fluorobenzeneboronic acid is used as a reactant for Rh-catalyzed asymmetric addition reactions, palladium-catalyzed oxidative cross-coupling reaction and Suzuki-Miyaura coupling.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144432-85-9
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD00051800
InChI Key WJDZZXIDQYKVDG-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
PubChem CID 2734660
IUPAC Name (3-chloro-4-fluorophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)Cl)(O)O

Specifications

Melting Point 255°C to 260°C
Quantity 25 g
Beilstein 7369789
Formula Weight 174.37
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.