missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenol, 99%, Thermo Scientific™

Product Code. 10252022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10252022 5 g Glass bottle 5g
10491481 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10252022 Supplier Thermo Scientific Acros Supplier No. 277200050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2613-23-2
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.55
MDL Number MFCD00002257
InChI Key ZQXLIXHVJVAPLW-UHFFFAOYSA-N
Synonym phenol, 3-chloro-4-fluoro, 3-chloro-4-fluoro-phenol, pubchem1492, pubchem2856, 3-chloro-4-fluorophe, 3-chloro4-fluorophenol, acmc-209goi, 4-fluoro-3-chlorophenol, 3-chloro-4-fluorophenol, ksc204s3j
PubChem CID 75790
IUPAC Name 3-chloro-4-fluorophenol
SMILES C1=CC(=C(C=C1O)Cl)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 39°C to 44°C
Color Beige to Brown
Boiling Point 104°C (11.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H3FOH
Quantity 5 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 146.55
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 3-Chloro-4-fluorophenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-4-fluorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.