missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenyl isocyanate, 98%

Product Code. 11343056
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343056 25 g 25g
11323056 1 g 1g
11333056 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343056 Supplier Thermo Scientific Alfa Aesar Supplier No. L10891.14
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 50529-33-4 | C7H3ClFNO | 171.555 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50529-33-4
Molecular Formula C7H3ClFNO
Molecular Weight (g/mol) 171.555
MDL Number MFCD00037037
InChI Key XVIPJBUXMFLHSI-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorophenyl isocyanate, 3-chloro-4-fluorophenylisocyanate, benzene, 2-chloro-1-fluoro-4-isocyanato, 3-chloro-4-fluorobenzenisocyanate, pubchem5024, acmc-20an6r, 3-chloro-4-fluoro-phenylisocyanate, 3-chloro-4-fluorophenyl-isocyanate, 3-chloro-4-fluoro phenyl isocyanate
PubChem CID 2734632
IUPAC Name 2-chloro-1-fluoro-4-isocyanatobenzene
SMILES C1=CC(=C(C=C1N=C=O)Cl)F

Specifications

Density 1.377
Boiling Point 44°C (17 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.541
Quantity 25 g
UN Number UN2206
Beilstein 973324
Sensitivity Moisture sensitive
Formula Weight 171.56
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.