missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenylboronic acid, 97%, ACROS Organics™

Product Code. 10447212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10447212 1 g Glass bottle 1g
10560542 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10447212 Supplier Thermo Scientific Acros Supplier No. 367680010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 144432-85-9
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD00051800
InChI Key WJDZZXIDQYKVDG-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
PubChem CID 2734660
IUPAC Name (3-chloro-4-fluorophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)Cl)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 174.37
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 3-Chloro-4-fluorophenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.