missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Product Code. 10515680 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 10515680

Brand: Thermo Scientific Acros 431990500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 413589-34-1 | C6H3BrClFMg | 233.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 413589-34-1, 109-99-9
CAS Min % 9.68, 85.48
CAS Max % 14.52, 90.32
Color Yellow to Brown
Density 0.9660g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C6H3BrClFMg
MDL Number MFCD02260212
Quantity 50 mL
Specific Gravity 0.966
Synonym bromo 3-chloro-4-fluorophenyl magnesium, 3-chloro-4-fluorophenylmagnesium bromide, xrncdmqvkmytam-uhfffaoysa-m, 3-chloro-4-fluorophenyl magnesium bromide, magnesium,1-chloro-2-fluorobenzene-5-ide,bromide, 3-chloro-4-fluorophenylmagnesium bromide, 0.50 m in 2-methf, 3-chloro-4-fluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3-chloro-4-fluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
InChI Key XRNCDMQVKMYTAM-UHFFFAOYSA-M
SMILES FC1=CC=C([Mg]Br)C=C1Cl
IUPAC Name magnesium(2+) 3-chloro-4-fluorobenzen-1-ide bromide
Molecular Weight (g/mol) 233.75
PubChem CID 2778256
Formula Weight 233.75
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluorophenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Chloro-4-fluorophenylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.