missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorothiophenol, 97%

Product Code. 11449832
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449832 1 g 1g
11459832 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449832 Supplier Thermo Scientific Alfa Aesar Supplier No. A10856.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 60811-23-6 | C6H3ClFS | 161.60 g/mol

Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharamaceutical intermediate.

Solubility
Sparingly Soluble (0.014 g/L) (25°C).

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60811-23-6
Molecular Formula C6H3ClFS
Molecular Weight (g/mol) 161.60
MDL Number MFCD00052938
InChI Key SFSHSIFYTWIGSF-UHFFFAOYSA-M
Synonym 3-chloro-4-fluorothiophenol, 3-chloro-4-fluorobenzene-1-thiol, 3-chloro-4-fluoro-benzenethiol, pubchem2857, acmc-209wz2, 3-chloro-4-flurothiophenol, benzenethiol, 3-chloro-4-fluoro
PubChem CID 2734197
IUPAC Name 3-chloro-4-fluorobenzenethiol
SMILES FC1=CC=C([S-])C=C1Cl

Specifications

Boiling Point 68°C to 70°C (2 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble (0.014g/L) (25°C).
Formula Weight 162.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-fluorothiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.