missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxyacetanilide, 98%

Product Code. 15249305 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15249305

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3964-54-3 | C8H8ClNO2 | 185.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3964-54-3
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD01318123
InChI Key JULKJDRBSRRBHT-UHFFFAOYSA-N
Synonym n-3-chloro-4-hydroxyphenyl acetamide, 3-chloro-4-hydroxyacetanilide, 4-acetylamino-2-chlorophenol, 2-chloro-4-acetamidophenol, acetamide, n-3-chloro-4-hydroxyphenyl, 3'-chloro-4'-hydroxyacetanilide, zlchem 1169, paracetamol impurity c, chloro-4-acetamidophenol, 3-chloro-4-hydroxyacet anilide
PubChem CID 77579
IUPAC Name N-(3-chloro-4-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=C(O)C(Cl)=C1

Specifications

Melting Point 144.0°C to 146.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in hot water
Formula Weight 185.61
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-hydroxyacetanilide
compliance-icons
Product Identifier
  • 3-Chloro-4-hydroxyacetanilide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.