missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxybenzonitrile, 95%

Product Code. p-7033338
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11315469 1 g 1g
11325469 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11315469 Supplier Thermo Scientific Alfa Aesar Supplier No. H50434.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2315-81-3 | C7H4ClNO | 153.565 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2315-81-3
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.565
MDL Number MFCD01567246
InChI Key CRYPJUOSZDQWJZ-UHFFFAOYSA-N
Synonym benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94
PubChem CID 2735739
IUPAC Name 3-chloro-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1C#N)Cl)O

Specifications

Melting Point 147°C to 149°C
Quantity 1 g
Formula Weight 153.57
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-hydroxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.