missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methoxybenzoic acid, 98+%

Product Code. 11419342
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419342 5 g 5g
11429342 25 g 25g
11439342 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419342 Supplier Thermo Scientific Alfa Aesar Supplier No. A10106.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37908-96-6 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37908-96-6
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00016512
InChI Key IBCQUQXCTOPJOD-UHFFFAOYSA-N
Synonym 3-chloro-p-anisic acid, 3-chloro-4-methoxybenzolic acid, benzoic acid, 3-chloro-4-methoxy, pubchem19130, acmc-209iw4, ksc494s0j, 3-chloro-4-methoxybenzoicacid, rarechem al bo 2400, 3-chloro-4-anisic acid, 3-chloro-4-methoxy benzoic acid
PubChem CID 169982
IUPAC Name 3-chloro-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)Cl

Specifications

Melting Point 215°C to 219°C
Quantity 5 g
Beilstein 2087584
Formula Weight 186.59
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-4-methoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.