missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylaniline, 98%

Product Code. 11472787
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472787 250 g 250g
11462787 1000 g 1000g
11329499 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472787 Supplier Thermo Scientific Alfa Aesar Supplier No. A15439.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 95-74-9 | C7H8ClN | 141.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-74-9
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007773
InChI Key RQKFYFNZSHWXAW-UHFFFAOYSA-N
Synonym 3-chloro-p-toluidine, 2-chloro-4-aminotoluene, 4-amino-2-chlorotoluene, benzenamine, 3-chloro-4-methyl, 3-chloro-4-methylbenzenamine, 3-chloro-4-methylphenylamine, 1-amino-3-chloro-4-methylbenzene, 4-methyl-3-chloroaniline, p-toluidine, 3-chloro, gull toxicant
PubChem CID 7255
ChEBI CHEBI:37824
IUPAC Name 3-chloro-4-methylaniline
SMILES CC1=C(C=C(C=C1)N)Cl

Specifications

Melting Point 22°C to 24°C
Density 1.18
Boiling Point 236°C to 238°C
Flash Point 130°C (266°F)
Refractive Index 1.584
Quantity 250 g
UN Number UN2239
Beilstein 636511
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.