missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylaniline, 98%

Product Code. p-7022590
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11472787

Brand: Thermo Scientific Alfa Aesar A15439.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-74-9 | C7H8ClN | 141.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-74-9
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007773
InChI Key RQKFYFNZSHWXAW-UHFFFAOYSA-N
Synonym 3-chloro-p-toluidine, 2-chloro-4-aminotoluene, 4-amino-2-chlorotoluene, benzenamine, 3-chloro-4-methyl, 3-chloro-4-methylbenzenamine, 3-chloro-4-methylphenylamine, 1-amino-3-chloro-4-methylbenzene, 4-methyl-3-chloroaniline, p-toluidine, 3-chloro, gull toxicant
PubChem CID 7255
ChEBI CHEBI:37824
IUPAC Name 3-chloro-4-methylaniline
SMILES CC1=C(C=C(C=C1)N)Cl

Specifications

Melting Point 22°C to 24°C
Density 1.18
Boiling Point 236°C to 238°C
Flash Point 130°C (266°F)
Refractive Index 1.584
Quantity 250 g
UN Number UN2239
Beilstein 636511
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-methylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.