missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethyl)benzoic acid, 97%

Product Code. 15444717
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444717 5 g 5g
15434717 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444717 Supplier Thermo Scientific Alfa Aesar Supplier No. H32038.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115754-20-6 | C8H4ClF3O2 | 224.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 115754-20-6
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD04972755
InChI Key UDXPRKSPAZWHQN-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethyl benzoic acid, benzoic acid, 3-chloro-4-trifluoromethyl, 3-chloro-4-trifluoromethyl-benzoic acid, acmc-1c5tn, chloro-4-trifluoromethyl benzoic acid, 3-chloro-4-trifluoromethyl benzoic acid, jrd
PubChem CID 21180297
IUPAC Name 3-chloro-4-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(Cl)=C(C=C1)C(F)(F)F

Especificaciones

Melting Point 158°C to 161°C
Quantity 5 g
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.